Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183349.1 | NP_899192.1 | 13 - 14 | 1637 | 106 | |
GenBank mRNA | AY313897.1 | - | 13 - 14 | 1637 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |