Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lztfl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199587.1 | NP_955881.1 | 8 - 9 | 937 | 101 | |
XM_005170618.4 | XP_005170675.1 | 8 - 9 | 983 | 101 | ||
GenBank mRNA | BC046036.1 | - | 8 - 9 | 937 | 101 | |
MK253678.1 | - | 7 - 8 | 773 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |