Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mpped2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200042.1 | NP_956336.1 | 5 - 6 | 1024 | 85 | |
XM_005174308.4 | XP_005174365.1 | 5 - 6 | 1030 | 85 | ||
GenBank mRNA | AY639016.1 | - | 4 - 5 | 768 | 85 | |
BC049341.1 | - | 5 - 6 | 1024 | 85 | ||
BC153488.1 | - | 5 - 6 | 1003 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |