Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nudt4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001310061.1 | NP_001296990.1 | 2 - 3 | 665 | 149 | |
NM_001310062.1 | NP_001296991.1 | 2 - 3 | 665 | 146 | ||
GenBank mRNA | AY423033.1 | - | 2 - 3 | 483 | 98 | |
BC091886.1 | - | 2 - 3 | 370 | 149 | ||
BC164969.1 | - | 2 - 3 | 248 | 149 | ||
EH442667.1 | - | 2 - 3 | 691 | 146 |
Set Membership: |
Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |