Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gabra6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200731.1 | NP_957025.1 | 7 - 8 | 896 | 95 | |
XM_005173112.4 | XP_005173169.1 | 7 - 8 | 1295 | 95 | ||
GenBank mRNA | BC059508.1 | - | 7 - 8 | 896 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |