Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dip2ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212963.1 | NP_998128.1 | 33 - 34 | 4190 | 91 | |
XM_009296840.3 | XP_009295115.1 | 35 - 36 | 4258 | 91 | ||
XM_009296841.3 | XP_009295116.1 | 34 - 35 | 4252 | 91 | ||
XM_009296842.3 | XP_009295117.1 | 34 - 35 | 4234 | 91 | ||
GenBank mRNA | BC068015.1 | - | 33 - 34 | 4190 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |