Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif3i | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212990.1 | NP_998155.1 | 4 - 5 | 402 | 105 | |
GenBank mRNA | BC045995.1 | - | 4 - 5 | 402 | 105 | |
BC065874.1 | - | 4 - 5 | 344 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |