Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tfr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009917.1 | NP_001009917.1 | 2 - 3 | 261 | 88 | |
XM_005163666.4 | XP_005163723.1 | 3 - 4 | 362 | 88 | ||
GenBank mRNA | AY649363.1 | - | 2 - 3 | 261 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |