Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asrgl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013529.1 | NP_001013547.1 | 6 - 7 | 821 | 73 | |
GenBank mRNA | BC090901.1 | - | 6 - 7 | 821 | 73 | |
BC164091.1 | - | 5 - 6 | 754 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |