Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: blk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030220.1 | NP_001025391.1 | 9 - 10 | 783 | 78 | |
XM_017352511.2 | XP_017208000.1 | 8 - 9 | 1383 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |