Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctdsplb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076615.1 | NP_001070083.1 | 5 - 6 | 808 | 97 | |
XM_005163361.4 | XP_005163418.1 | 6 - 7 | 1115 | 97 | ||
GenBank mRNA | BC124542.1 | - | 5 - 6 | 808 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |