Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jmjd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076628.1 | NP_001070096.1 | 5 - 6 | 1305 | 78 | |
GenBank mRNA | BC124503.1 | - | 5 - 6 | 1305 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |