Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130954.2 | NP_571029.2 | 7 - 8 | 1445 | 96 | |
GenBank mRNA | AF030031.1 | - | 7 - 8 | 1436 | 96 | |
BC075742.1 | - | 7 - 8 | 1445 | 96 | ||
CU458989.1 | - | 7 - 8 | 1436 | 96 | ||
DQ993185.1 | - | 7 - 8 | 1436 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |