Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: col2a1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131292.1 | NP_571367.1 | 51 - 52 | 3945 | 62 | |
XM_005166863.4 | XP_005166920.1 | 50 - 51 | 3877 | 62 | ||
GenBank mRNA | BC093433.1 | - | 1 - 2 | 204 | 62 | |
BC163934.1 | - | 51 - 52 | 3937 | 62 | ||
BC167477.1 | - | 50 - 51 | 3841 | 62 | ||
BC168504.1 | - | 51 - 52 | 4026 | 62 | ||
DQ335127.1 | - | 51 - 52 | 3945 | 62 | ||
U23822.1 | - | 8 - 9 | 820 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |