Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mif4gda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200384.1 | NP_956678.1 | 4 - 5 | 468 | 70 | |
XM_005166179.4 | XP_005166236.3 | 4 - 5 | 780 | 70 | ||
GenBank mRNA | BC053283.1 | - | 4 - 5 | 468 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |