Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fahd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212918.1 | NP_998083.1 | 3 - 4 | 601 | 98 | |
XM_021478226.1 | XP_021333901.1 | 3 - 4 | 513 | 98 | ||
XM_021478227.1 | XP_021333902.1 | 3 - 4 | 662 | 98 | ||
XM_021478228.1 | XP_021333903.1 | 3 - 4 | 513 | 98 | ||
XM_021478229.1 | XP_021333904.1 | 3 - 4 | 597 | 98 | ||
GenBank mRNA | BC067576.1 | - | 3 - 4 | 601 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |