Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cul1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213495.1 | NP_998660.1 | 9 - 10 | 1345 | 68 | |
XM_021470126.1 | XP_021325801.1 | 9 - 10 | 1432 | 68 | ||
GenBank mRNA | BC048370.1 | - | 9 - 10 | 1345 | 68 | |
BC165368.1 | - | 8 - 9 | 1111 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |