Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sec11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002521.2 | NP_001002521.1 | 4 - 5 | 543 | 121 | |
GenBank mRNA | BC076276.1 | - | 4 - 5 | 547 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |