Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sec11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002521.2 | NP_001002521.1 | 5 - 6 | 602 | 80 | |
GenBank mRNA | BC076276.1 | - | 5 - 6 | 606 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |