Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: olfm1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003486.2 | NP_001003486.1 | 1 - 2 | 448 | 67 | |
NM_001328415.1 | NP_001315344.1 | 1 - 2 | 448 | 67 | ||
GenBank mRNA | BC078378.1 | - | 1 - 2 | 325 | 67 | |
EH432606.1 | - | 1 - 2 | 472 | 67 | ||
EH493055.1 | - | 1 - 2 | 197 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |