Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wdr54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003630.2 | NP_001003630.2 | 6 - 7 | 814 | 89 | |
XM_005165298.4 | XP_005165355.1 | 6 - 7 | 842 | 89 | ||
GenBank mRNA | BC077087.1 | - | 6 - 7 | 818 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |