Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hibadha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003646.2 | NP_001003646.1 | 6 - 7 | 693 | 90 | |
GenBank mRNA | BC078207.1 | - | 6 - 7 | 696 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |