Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zfand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003651.1 | NP_001003651.1 | 1 - 2 | 62 | 73 | |
XM_021470141.1 | XP_021325816.1 | 1 - 2 | 78 | 73 | ||
GenBank mRNA | BC076430.1 | - | 1 - 2 | 62 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |