Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gbgt1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004620.1 | NP_001004620.1 | 1 - 2 | 209 | 71 | |
XM_021476097.1 | XP_021331772.1 | 1 - 2 | 215 | 71 | ||
GenBank mRNA | BC081406.1 | - | 1 - 2 | 209 | 71 | |
BC164153.1 | - | 1 - 2 | 90 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |