Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bmp2k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008644.2 | NP_001008644.2 | 2 - 3 | 654 | 91 | |
NM_001193540.1 | NP_001180469.1 | 2 - 3 | 654 | 91 | ||
XM_005165303.4 | XP_005165360.1 | 2 - 3 | 1299 | 91 | ||
GenBank mRNA | BC086712.1 | - | 2 - 3 | 654 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |