Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxo16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017680.1 | NP_001017680.1 | 7 - 8 | 993 | 106 | |
GenBank mRNA | BC093325.1 | - | 7 - 8 | 993 | 106 | |
BC164389.1 | - | 6 - 7 | 814 | 106 |
Gene Symbol: LOC100150082 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128401.1 | NP_001121873.1 | 7 - 8 | 1109 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |