Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scn4ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045065.1 | NP_001038530.1 | 3 - 4 | 476 | 114 | |
XM_005171585.4 | XP_005171642.1 | 4 - 5 | 827 | 114 | ||
GenBank mRNA | BC163558.1 | - | 3 - 4 | 522 | 114 | |
DQ149505.1 | - | 3 - 4 | 476 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |