Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rwdd2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075115.1 | NP_001068583.1 | 3 - 4 | 381 | 65 | |
GenBank mRNA | BC124264.1 | - | 3 - 4 | 381 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |