Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sox17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131287.2 | NP_571362.2 | 1 - 2 | 444 | 85 | |
GenBank mRNA | AF168614.2 | - | 1 - 2 | 419 | 85 | |
BC086959.1 | - | 1 - 2 | 444 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |