Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mmgt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205599.1 | NP_991162.1 | 3 - 4 | 294 | 77 | |
GenBank mRNA | AY394962.1 | - | 3 - 4 | 294 | 77 | |
BC076369.1 | - | 3 - 4 | 282 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |