Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nf2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212951.1 | NP_998116.1 | 7 - 8 | 825 | 134 | |
GenBank mRNA | AY561520.1 | - | 7 - 8 | 825 | 134 | |
BC079503.1 | - | 7 - 8 | 762 | 134 | ||
BC108005.1 | - | 7 - 8 | 831 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |