Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdk5rap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002105.1 | NP_001002105.1 | 9 - 10 | 1004 | 123 | |
GenBank mRNA | BC071504.1 | - | 9 - 10 | 1004 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |