Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acp5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002452.1 | NP_001002452.1 | 4 - 5 | 724 | 62 | |
GenBank mRNA | BC076019.1 | - | 4 - 5 | 724 | 62 | |
HM239641.1 | - | 4 - 5 | 556 | 62 | ||
HM239642.1 | - | 4 - 5 | 555 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |