Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnd3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002591.1 | NP_001002591.1 | 4 - 5 | 613 | 98 | |
GenBank mRNA | AY865560.1 | - | 4 - 5 | 439 | 98 | |
BC076009.1 | - | 4 - 5 | 613 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |