Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pkmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003488.1 | NP_001003488.1 | 4 - 5 | 576 | 75 | |
XM_005163113.4 | XP_005163170.3 | 5 - 6 | 1084 | 75 | ||
GenBank mRNA | BC076497.1 | - | 4 - 5 | 576 | 75 | |
BC165710.1 | - | 4 - 5 | 586 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |