Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trpa1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007065.1 | NP_001007066.1 | 8 - 9 | 972 | 122 | |
XM_017358267.2 | XP_017213756.1 | 7 - 8 | 967 | 122 | ||
GenBank mRNA | AY677196.1 | - | 8 - 9 | 972 | 122 | |
EU826643.1 | - | 7 - 8 | 941 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |