Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mtmr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014330.2 | NP_001014352.1 | 14 - 15 | 1893 | 95 | |
XM_005161411.4 | XP_005161468.1 | 13 - 14 | 1895 | 95 | ||
GenBank mRNA | BC091956.1 | - | 14 - 15 | 1897 | 95 | |
BC165859.1 | - | 14 - 15 | 1660 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |