Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: neurl1ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076708.1 | NP_001070176.2 | 2 - 3 | 385 | 88 | |
GenBank mRNA | BC124268.1 | - | 2 - 3 | 385 | 88 | |
BC164216.1 | - | 1 - 2 | 201 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |