Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: comtb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083843.1 | NP_001077312.1 | 3 - 4 | 512 | 71 | |
GenBank mRNA | BC134931.1 | - | 3 - 4 | 512 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |