Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: coq2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089486.1 | NP_001082955.1 | 4 - 5 | 706 | 92 | |
GenBank mRNA | BC134043.1 | - | 4 - 5 | 706 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |