Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snai1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313699.1 | NP_001300628.1 | 2 - 3 | 313 | 63 | |
NM_001313700.1 | NP_001300629.1 | 1 - 2 | 297 | 63 | ||
GenBank mRNA | BC049300.1 | - | 1 - 2 | 222 | 63 | |
BC092698.1 | - | 1 - 2 | 127 | 63 | ||
BC152602.1 | - | 2 - 3 | 314 | 63 | ||
BC164590.1 | - | 1 - 2 | 115 | 63 | ||
EH544887.1 | - | 1 - 2 | 300 | 63 | ||
S68799.1 | - | 1 - 2 | 298 | 63 | ||
X74790.1 | - | 195 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |