Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpeb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131427.2 | NP_571502.2 | 9 - 10 | 1430 | 64 | |
XM_021477394.1 | XP_021333069.1 | 9 - 10 | 1444 | 64 | ||
GenBank mRNA | AB011680.1 | - | 8 - 9 | 1394 | 64 | |
AF076918.1 | - | 9 - 10 | 1409 | 64 | ||
BC045467.1 | - | 9 - 10 | 1428 | 64 | ||
BC164038.1 | - | 9 - 10 | 1428 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |