Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elavl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244600.1 | NP_001231529.1 | 4 - 5 | 970 | 101 | |
NM_001244601.1 | NP_001231530.1 | 4 - 5 | 844 | 101 | ||
NM_131453.3 | NP_571528.2 | 4 - 5 | 605 | 101 | ||
XM_005166786.4 | XP_005166843.1 | 3 - 4 | 882 | 101 | ||
XM_017357272.2 | XP_017212761.1 | 3 - 4 | 721 | 101 | ||
XM_021478178.1 | XP_021333853.1 | 4 - 5 | 968 | 101 | ||
XM_021478179.1 | XP_021333854.1 | 4 - 5 | 968 | 101 | ||
XM_021478180.1 | XP_021333855.1 | 4 - 5 | 968 | 101 | ||
GenBank mRNA | BC065965.1 | - | 4 - 5 | 605 | 101 | |
U17602.1 | - | 3 - 4 | 451 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |