Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp6v1ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182878.1 | NP_878298.1 | 1 - 2 | 205 | 83 | |
GenBank mRNA | AF472614.1 | - | 1 - 2 | 205 | 83 | |
BC052222.1 | - | 1 - 2 | 163 | 83 | ||
BC059455.1 | - | 1 - 2 | 155 | 83 | ||
BC071387.1 | - | 1 - 2 | 203 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |