Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp6v1ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182878.1 | NP_878298.1 | 3 - 4 | 359 | 60 | |
GenBank mRNA | AF472614.1 | - | 3 - 4 | 359 | 60 | |
BC052222.1 | - | 3 - 4 | 317 | 60 | ||
BC059455.1 | - | 3 - 4 | 309 | 60 | ||
BC071387.1 | - | 3 - 4 | 357 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |