Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aldoaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194377.2 | NP_919358.2 | 5 - 6 | 916 | 71 | |
XM_005164116.4 | XP_005164173.1 | 5 - 6 | 962 | 71 | ||
GenBank mRNA | AY394966.1 | - | 5 - 6 | 916 | 71 | |
BC044379.1 | - | 5 - 6 | 895 | 71 | ||
BC065320.1 | - | 5 - 6 | 866 | 71 | ||
BC164192.1 | - | 4 - 5 | 835 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |