Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tpst2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200419.1 | NP_956713.1 | 2 - 3 | 493 | 70 | |
XM_021479120.1 | XP_021334795.1 | 1 - 2 | 559 | 70 | ||
GenBank mRNA | BC054637.1 | - | 2 - 3 | 493 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |