Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: macrod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200549.1 | NP_956843.1 | 13 - 14 | 970 | 77 | |
XM_017358720.2 | XP_017214209.1 | 13 - 14 | 1096 | 77 | ||
GenBank mRNA | BC056529.1 | - | 13 - 14 | 970 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |