Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gorasp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200703.2 | NP_956997.1 | 1 - 2 | 184 | 75 | |
XM_021476811.1 | XP_021332486.1 | 1 - 2 | 208 | 75 | ||
GenBank mRNA | AY398354.1 | - | 1 - 2 | 184 | 75 | |
BC059447.1 | - | 1 - 2 | 145 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |