Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dldh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201506.1 | NP_958914.1 | 1 - 2 | 112 | 67 | |
GenBank mRNA | AY391421.1 | - | 1 - 2 | 63 | 67 | |
BC044432.1 | - | 1 - 2 | 112 | 67 | ||
BC065616.1 | - | 1 - 2 | 89 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |